DI Liner Adapter for Agilent 5890 Packed Column GC, Restek
Supplier: Restek
21303
RK21303KT 183.99 USD
RK21303
DI Liner Adapter for Agilent 5890 Packed Column GC, Restek
Chromatography Supplies Chromatography Spare Parts
Direct injection liner adapters use standard 1/16-inch capillary nuts and ferrules.
  • Convenient wrench pad at base
  • Includes 1/4-inch graphite ferrule and stainless steel nut
Use with Agilent or Restek DI glass inserts or Restek DI Uniliner® liners for an Agilent 5890 packed column GC.