Recessed Double Taper Inlet Liner for Agilent GCs equipped with split/splitless inlets, Restek
Supplier: Restek
20987
RK20987PK 257.85 USD
RK20987
Recessed Double Taper Inlet Liner for Agilent GCs equipped with split/splitless inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.