DI Uniliner® Liners for Agilent 5890 Packed Column GC, Restek
Supplier: Restek
20965
RK20965PK 152.82 USD
RK20965
DI Uniliner® Liners for Agilent 5890 Packed Column GC, Restek
Chromatography Supplies Chromatography Inlet Liners
Direct injection Uniliner® liners minimize solvent and peak tailing.
  • Press-Tight® taper forms dead volume free connection to column
  • Can be removed from the septum nut weldment
Fits same liner adapter as Agilent part #5181-3382 or #5080-8732.