Cup Inlet Liner with Top Taper for Thermo TRACE, 8000 Series and Focus GCs Equipped with SSL Inlets, Restek
Supplier: Restek
20951
RK20951PK 305.71 USD
RK20951
Cup Inlet Liner with Top Taper for Thermo TRACE, 8000 Series and Focus GCs Equipped with SSL Inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.