Uniliner® Inlet Liner with Wool for Agilent GCs equipped with split/splitless inlets, Restek
Supplier: Restek
22273
RK20844PK 423.04 USD
RK20844
Uniliner® Inlet Liner with Wool for Agilent GCs equipped with split/splitless inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.