Top Taper Inlet Liner for Bruker/Varian GCs equipped with 1075/1077 inlets, Restek
Supplier: Restek
20722
RK20722PK 185.17 USD
RK20722 75779-108
Top Taper Inlet Liner for Bruker/Varian GCs equipped with 1075/1077 inlets, Restek
Chromatography Supplies Chromatography Inlet Liners
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.