MSD Source Nut, Restek
Supplier: Restek
20643
RK20643PK 49.14 USD
RK20643
MSD Source Nut, Restek
Fittings and Connectors
The design of these nuts enhances ease of threading onto the transfer line and improves overall lifetime.
  • 1.2 mm nut bore permits easy removal of ferrules with a standard tapered-needle file
  • Made of brass to prevent thread-stripping on the transfer line