Polyimide, resin

Supplier: RESTEK CORP

20445
RK20445EA 60.1 USD
RK20445
Polyimide, resin
Polyimide

Resin securely connects a Press-Tight® connector to a fused silica column.


  • Max operating temperature of 350°C
  • Polyimide

Formula: C₃₅H₂₈N₂O₇
MDL Number: MFCD01868102
CAS Number: 62929-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR