Tubing Cutter, Restek
Supplier: Restek
20192
RK20192EA 155.33 USD
RK20192
Tubing Cutter, Restek
Cutters
Produces square, smooth cuts in tubing.
  • Eliminates tubing distortion
  • Replaceable cutting wheel
Cuts hard or soft copper, aluminum, brass, stainless steel, Monel® alloy, or titanium. Efficient tubing cutter consistently cut lengths of tubing.