FID/Injector Cleaning Kit, Restek
Supplier: Restek
20120
RK20120EA 102.27 USD
RK20120
FID/Injector Cleaning Kit, Restek
Chromatography Supplies Chromatography Spare Parts
The FID/Injector cleaning kit is compatible with any injection port, allowing universal applicability.
  • Stainless steel cleaning tools
Delivery information: Kits include: Nylon tube brushes; pipe cleaner; stainless steel tube brushes; stainless steel surface brush; stainless steel jet reamers; and emery cloth.