Lead(II) acetate 10% (w/v) in 2% acetic acid

Supplier: Ricca Chemical

Synonyms: Acetic acid Lead(II) salt

RSOD2000-1A
RCSOD20-32EA 600 USD
RCSOD20-32
Lead(II) acetate 10% (w/v) in 2% acetic acid
Lead (II) acetate
Formula: (H₃CCOO)₂Pb
MW: 325.29 g/mol
MDL Number: MFCD00012452
CAS Number: 301-04-2
UN: 1616
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR