Sulfuric acid ≥87% (w/w)

Supplier: Ricca Chemical

RSD81000-2.5D
RCSD81-5PTCS 462 USD
RCSD81-5PT
Sulfuric acid ≥87% (w/w)
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR