Phenolphthalein 0,5% in methanol 80%
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
RSD54000-4A
RSD54000-1A
RCSD54-1CS
378
USD
RCSD54-1
RCSD54-32
Phenolphthalein 0,5% in methanol 80%
Phenolphthalein
Formula:
C₂₀H₁₄O₄ Boiling Pt: ~ 65 °C (Methanol) Density: 0.8345 g/cm³ Flash Pt: ~ 11 °C (MeOH) Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 UN: 1993 ADR: 3,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...