Bromothymol blue 0.04% in isopropanol (IPA)

Supplier: Ricca Chemical
Danger

RSD13000-500A
RCSD13-16EA 184 USD
RCSD13-16
Bromothymol blue 0.04% in isopropanol (IPA)
Bromothymol blue
Formula: C₂₇H₂₈Br₂O₅S
Boiling Pt: 82.5 °C [DIN 43171]
Melting Pt: –90 °C
Density: 0.7850 g/cm³
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
UN: 1219
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR