VeriSpec® Standard for ASTM D5580, RICCA Chemical Company
Supplier: Ricca Chemical
RV019034-1R
RCRV019034-1REA 357 USD
RCRV019034-1R
VeriSpec® Standard for ASTM D5580, RICCA Chemical Company
Standards Petroleum Industry Standards
Two components standard in in methanol.
2000 ppm: 2-bromo-1-chloropropane, fluorobenzene.
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility