VeriSpec® ICP Multi-Element Standard 29, RICCA Chemical Company
Supplier: Ricca Chemical
RV010860-100N
RCRV010860100NEA 865 USD
RCRV010860100N
VeriSpec® ICP Multi-Element Standard 29, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
29 component standard.
10 ppm: Al, Ag, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Ti, U, V, Zn.
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility