VeriSpec® Trace Metals Standard I, RICCA Chemical Company
Supplier: Ricca Chemical
RV010852-100N
RCRV010852100NEA 543 USD
RCRV010852100N
VeriSpec® Trace Metals Standard I, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
15 components standard.
10 ppm: Hg
25 ppm: Cd, Se
100 ppm: As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn
250 ppm: V
500 ppm: Al
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility