VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Supplier: Ricca Chemical
RV010741-100N
RCRV010741100NEA
463
USD
RCRV010741100N
VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Standards
Anionic Standards
Multi-Element Anioinic Standards
Certified reference material (CRM) for ion chromatography (IC)
100 ppm: F-, PO₄³⁺, NO₂-, NO₃-, Br-, SO₄²⁺, Cl-
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)