VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Supplier: Ricca Chemical
RV010738-100N
RCRV010738100NEA 461 USD
RCRV010738100N
VeriSpec® Mixed Anion Standard, 7 Component, Ricca Chemical Company
Standards Anionic Standards Multi-Element Anioinic Standards
Certified reference material (CRM) for ion chromatography (IC)
2 ppm F-, 10 ppm PO₄³⁺, NO₃-, Br-, SO₄²⁺, 5ppm Cl-, NO₂-
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility