VeriSpec™ As⁵⁺ Standard for ICP 100 ppm
Supplier: Ricca Chemical
RV010640-100N
RCRV010640100NEA 180 USD
RCRV010640100N
VeriSpec™ As⁵⁺ Standard for ICP 100 ppm
Standards Cationic Standards Arsenic Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility