VeriSpec® Tungsten Standard for ICP/MS 10 ppm NH₄OH, RICCA Chemical Company
Supplier: Ricca Chemical
RV010634-100N
RCRV010634100NEA 140 USD
RCRV010634100N
VeriSpec® Tungsten Standard for ICP/MS 10 ppm NH₄OH, RICCA Chemical Company
Standards Cationic Standards Tungsten Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility