VeriSpec® Uranium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010632-50N
RV010632-100N
RCRV010632-50NEA
86.45
USD
RCRV010632-50N
RCRV010632100N
VeriSpec® Uranium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Uranium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)