VeriSpec® Tin Standard for ICP/MS 10 ppm in 1% HNO₃/HF, RICCA Chemical Company
Supplier: Ricca Chemical
RV010623-100N
RCRV010623-50NEA 86.97 USD
RCRV010623-50N RCRV010623100N
VeriSpec® Tin Standard for ICP/MS 10 ppm in 1% HNO₃/HF, RICCA Chemical Company
Standards Cationic Standards Tin Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility