VeriSpec® Silicon Standard for ICP/MS 10 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010621-100N
RCRV010621100NEA 143 USD
RCRV010621100N
VeriSpec® Silicon Standard for ICP/MS 10 ppm in H₂O, RICCA Chemical Company
Standards Cationic Standards Silicium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility