VeriSpec® Rhenium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010614-100N
RCRV010614100NEA
369
USD
RCRV010614100N
VeriSpec® Rhenium Standard for ICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Standards
Cationic Standards
Rhenium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)