VeriSpec™ Praseodymium Standard forICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010611-100N
RCRV010611100NEA 140 USD
RCRV010611100N
VeriSpec™ Praseodymium Standard forICP/MS 10 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Praseodymium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility