VeriSpec™ Niobium Standard for ICP/MS 10 ppm in 2% HNO₃/HF, RICCA Chemical Company
Supplier: Ricca Chemical
RV010605-100N
RCRV010605100NEA
138
USD
RCRV010605100N
VeriSpec™ Niobium Standard for ICP/MS 10 ppm in 2% HNO₃/HF, RICCA Chemical Company
Standards
Cationic Standards
Niobium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)