VeriSpec® Holmium Standard for ICP/MS 10 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010594-100N
RCRV010594-50NEA 80.82 USD
RCRV010594-50N RCRV010594100N
VeriSpec® Holmium Standard for ICP/MS 10 ppm in 2% HNO₃
Standards Cationic Standards Holmium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility