VeriSpec™ Hafnium Standard for ICP/MS 10 ppm in 2% HNO?/HF
Supplier: Ricca Chemical
RV010592-100N
RCRV010592100NEA 259 USD
RCRV010592100N
VeriSpec™ Hafnium Standard for ICP/MS 10 ppm in 2% HNO?/HF
Standards Cationic Standards Hafnium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility