VeriSpec® Vanadium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010564-100N
RCRV010564100NEA 149 USD
RCRV010564100N
VeriSpec® Vanadium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Vanadium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility