VeriSpec® Uranium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010563-100N
RCRV010563100NEA 207 USD
RCRV010563100N
VeriSpec® Uranium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Uranium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility