VeriSpec® Rubidium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010545-100N
RCRV010545100NEA 159 USD
RCRV010545100N
VeriSpec® Rubidium Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Rubidium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility