VeriSpec™ Iridium Standard for ICP 100 ppm in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010528-100N
RCRV010528100NEA 289 USD
RCRV010528100N
VeriSpec™ Iridium Standard for ICP 100 ppm in 2% HCl, RICCA Chemical Company
Standards Cationic Standards Iridium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility