VeriSpec® Indium Standard for ICP/MS 100 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010527-100N
RCRV010527100NEA 173 USD
RCRV010527100N
VeriSpec® Indium Standard for ICP/MS 100 ppm in 2% HNO₃
Standards Cationic Standards Indium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility