VeriSpec® Holmium Standard for ICP/MS 100 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010526-100N
RCRV010526100NEA 159 USD
RCRV010526100N
VeriSpec® Holmium Standard for ICP/MS 100 ppm in 2% HNO₃
Standards Cationic Standards Holmium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility