VeriSpec® Copper Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010516-100N
RCRV010516100NEA 153 USD
RCRV010516100N
VeriSpec® Copper Standard for ICP/MS 100 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Copper Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility