VeriSpec® Strontium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010488-100N
RCRV010488-50NEA 71.65 USD
RCRV010488-50N RCRV010488100N RCRV010488250N RCRV010488500N
VeriSpec® Strontium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Strontium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility