VeriSpec® Rubidium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010477-100N
RCRV010477-50NEA 172.07 USD
RCRV010477-50N RCRV010477100N RCRV010477250N RCRV010477500N
VeriSpec® Rubidium Standard for ICP 10 g/L in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Rubidium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 10 g/L= 10,000 µg/mL