Beryllium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010439-100N
RCRV010439-50NEA 329.55 USD
RCRV010439-50N RCRV010439100N RCRV010439250N RCRV010439500N
Beryllium Standards, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Beryllium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml