Ytterbium Standard, for ICP, VeriSpec®, RICCA Chemical Company
Supplier: Ricca Chemical
RV010430-100N
RCRV010430100NEA 259 USD
RCRV010430100N RCRV010430250N RCRV010430500N
Ytterbium Standard, for ICP, VeriSpec®, RICCA Chemical Company
Standards Cationic Standards Ytterbium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml