Uranium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010426-100N
RCRV010426100NEA 409 USD
RCRV010426100N RCRV010426250N RCRV010426500N
Uranium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Uranium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml