Thallium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010424-100N
RCRV010424100NEA 239 USD
RCRV010424100N RCRV010424250N RCRV010424500N
Thallium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Thallium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml