VeriSpec® Silicon Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010415-500N
RCRV010415100NEA 225.01 USD
RCRV010415100N RCRV010415250N RCRV010415500N
VeriSpec® Silicon Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Standards Cationic Standards Silicium Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility