VeriSpec® Sulfur Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010411-100N
RV010411-250N
RV010411-500N
RCRV010411100NEA
259
USD
RCRV010411100N
RCRV010411250N
RCRV010411500N
VeriSpec® Sulfur Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Standards
Cationic Standards
Sulfur standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility