Cadmium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Supplier: Ricca Chemical
RV010372-100N
RCRV010372100NEA 239 USD
RCRV010372100N RCRV010372250N RCRV010372500N
Cadmium Standard, for ICP, VeriSpec®, Ricca Chemical Company
Standards Cationic Standards Cadmium Standards
  • NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml