VeriSpec® Boron Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010367-100N
RCRV010367100NEA 968 USD
RCRV010367100N RCRV010367250N RCRV010367500N
VeriSpec® Boron Standard for ICP 10 g/L in H₂O, RICCA Chemical Company
Standards Cationic Standards Boron Standards
Certified reference material (CRM) for ICP
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility