Nickel (II) nitrate 10000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy

Supplier: Ricca Chemical
Danger

Synonyms: Nickel dinitrate

RV010133-50N
RCRV010133-50NEA 269 USD
RCRV010133-50N
Nickel (II) nitrate 10000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Nickel (II) nitrate

Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Formula: N₂NiO₆
MW: 182.7 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011139
CAS Number: 13138-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR