Ammonium nitrate 50000 ppm in water, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy

Supplier: Ricca Chemical

RV010132-50N
RCRV010132-50NEA 419 USD
RCRV010132-50N
Ammonium nitrate 50000 ppm in water, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Ammonium nitrate

Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Formula: NH₄NO₃
MDL Number: MFCD00011425
CAS Number: 6484-52-2
UN: 1942
ADR: 5.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR