Ammonium dihydrogen phosphate 100,000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate
RV010131-50N
RCRV010131-50NEA
889
USD
RCRV010131-50N
RCRV010131100N
Ammonium dihydrogen phosphate 100,000 ppm in nitric acid 1%, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Ammonium dihydrogen phosphate
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility
Formula:
NH₄H₂PO₄ MW: 115.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003396 CAS Number: 7722-76-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...