VeriSpec™ Antimony (Sb) Standard for AAS 20 ppb in 1% HNO₃
Supplier: Ricca Chemical
RV010127-50N
RCRV010127-50NEA 239 USD
RCRV010127-50N
VeriSpec™ Antimony (Sb) Standard for AAS 20 ppb in 1% HNO₃
Standards Cationic Standards Antimony Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility